missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione Mono-2,2-dimethyltrimethylene Ketal 98.0+%, TCI America™

Catalog No. C23285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23285G 5 g
C232825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23285G Supplier TCI America Supplier No. C23285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69225-59-8
Molecular Formula C11H18O3
Molecular Weight (g/mol) 198.262
MDL Number MFCD00006652
InChI Key COKVDTKAWIFNTH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
PubChem CID 587968
IUPAC Name 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
SMILES CC1(COC2(CCC(=O)CC2)OC1)C

Specifications

Melting Point 51°C
Color White
Quantity 5 g
Formula Weight 198.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione Mono-2,2-dimethyltrimethylene Ketal