accessibility menu, dialog, popup

UserName

H3CH3COOO

CAS RN 69225-59-8

IUPAC Name: 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Synonyms: 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Molecular Weight (g/mol): 198.26
Molecular Formula: C11H18O3
InChi Key: COKVDTKAWIFNTH-UHFFFAOYSA-N
SMILES: CC1(C)COC2(CCC(=O)CC2)OC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results