missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexadiene (stabilized with BHT) 98.0+%, TCI America™

Supplier:  TCI America C046810ML

 View more versions of this product

Catalog No. C046810ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,4-Cyclohexadiene (stabilized with BHT)
-49°C
88°C
MFCD00001535
3295
UVJHQYIOXKWHFD-UHFFFAOYSA-N
cyclohexa-1,4-diene
12343
80.13
Liquid
628-41-1
Colorless
C6H8
10 mL
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
C1C=CCC=C1
80.13
CHEBI:37611
≥98.0% (GC)

Chemical Identifiers

628-41-1
80.13
UVJHQYIOXKWHFD-UHFFFAOYSA-N
12343
cyclohexa-1,4-diene
C6H8
MFCD00001535
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
CHEBI:37611
C1C=CCC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions