missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexadiene (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7124443
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C046810ML 10 mL
C046825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C046810ML Supplier TCI America Supplier No. C046810ML
Only null left

Chemical Identifiers

CAS 628-41-1
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001535
InChI Key UVJHQYIOXKWHFD-UHFFFAOYSA-N
Synonym 1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
PubChem CID 12343
ChEBI CHEBI:37611
IUPAC Name cyclohexa-1,4-diene
SMILES C1C=CCC=C1

Specifications

Melting Point -49°C
Color Colorless
Boiling Point 88°C
Quantity 10 mL
UN Number 3295
Formula Weight 80.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Cyclohexadiene (stabilized with BHT)
TSCA Yes
Recommended Storage Refrigerator