missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole, 97%, Thermo Scientific™

Catalog No. p-2694102
Change view
Click to view available options
Quantity:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB07116EA 10g
BTB07116EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB07116EA Supplier Thermo Scientific Chemicals Supplier No. BTB07116EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16851-83-5
Molecular Formula C10H8ClNO2S
Molecular Weight (g/mol) 241.689
MDL Number MFCD00067753
InChI Key ACNNPPGRQRFTSO-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl sulfonyl-1h-pyrrole, 1-4-chlorobenzenesulfonyl pyrrole, maybridge1_001769, cbmicro_045913, 1-4-chlorophenyl sulfonylpyrrole, 1-chloro-4-pyrrolylsulfonyl benzene, 4-chloro-1-pyrrolylsulfonyl benzene, 1-4-chlorobenzenesulfonyl-1h-pyrrole, 1-4-chlorophenylsulfonyl-1h-pyrrole
PubChem CID 706366
IUPAC Name 1-(4-chlorophenyl)sulfonylpyrrole
SMILES C1=CN(C=C1)S(=O)(=O)C2=CC=C(C=C2)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10g
Formula Weight 241.7
Percent Purity 97%
Chemical Name or Material 1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.