missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)cyclobutanecarbonitrile 98.0+%, TCI America™

Catalog No. p-7125864
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31615G 5 g
C316125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31615G Supplier TCI America Supplier No. C31615G

Chemical Identifiers

CAS 28049-61-8
Molecular Formula C11H10ClN
Molecular Weight (g/mol) 191.66
MDL Number MFCD00065239
InChI Key XQONXPWVIZZJIL-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclobutanecarbonitrile, 1-4-chlorophenyl-1-cyclobutanecarbonitrile, 1-4-chlorophenyl cyclobutane-1-carbonitrile, cyclobutanecarbonitrile, 1-4-chlorophenyl, 1-4-chlorophenyl-cyclobutanecarbonitrile, ccbc, sibutramine impurity f, acmc-20a2wi, intermediate of sibutramine
PubChem CID 98621
IUPAC Name 1-(4-chlorophenyl)cyclobutane-1-carbonitrile
SMILES ClC1=CC=C(C=C1)C1(CCC1)C#N

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 112°C
Formula Weight 191.66
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-(4-Chlorophenyl)cyclobutanecarbonitrile