accessibility menu, dialog, popup

UserName

CAS RN 28049-61-8

IUPAC Name: 1-(4-chlorophenyl)cyclobutane-1-carbonitrile
Synonyms: 1-(4-chlorophenyl)cyclobutane-1-carbonitrile
Molecular Weight (g/mol): 191.66
Molecular Formula: C11H10ClN
InChi Key: XQONXPWVIZZJIL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1(CCC1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results