missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)biguanide hydrochloride, 97%

Catalog No. AAB2031106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2031106 5 g
AAB2031103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2031106 Supplier Thermo Scientific Chemicals Supplier No. B2031106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4022-81-5 | C8H11Cl2N5 | 248.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4022-81-5
Molecular Formula C8H11Cl2N5
Molecular Weight (g/mol) 248.11
MDL Number MFCD00053020
InChI Key NAFSLMFLGYXGIF-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl biguanide hydrochloride, 1-p-chlorophenyl biguanide hydrochloride, 1-carbamimidamido-n-4-chlorophenyl methanimidamide hydrochloride, n-p-chlorophenyl biguanide hydrochloride, 4-chloroanilino imino methyl amino methanimidamide hydrochloride, n-4-chlorophenyl-imidocarbonimidic diamide monohydrochloride, 1-carbamimidoyl-3-4-chlorophenyl guanidine, acmc-20apaw, n-p-chlorophenylbiguanide hydrochloride, 1-4-chlorophenyl-biguanide hydrochloride
PubChem CID 458746
SMILES [Cl-].NC([NH3+])=NC(N)=NC1=CC=C(Cl)C=C1

Specifications

Melting Point 247°C to 250°C
Quantity 5 g
Beilstein 3732533
IUPAC Name 2-(4-chlorophenyl)-1-(diaminomethylidene)guanidine;hydrochloride
Formula Weight 248.12
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)biguanide hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber DU1780200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only