accessibility menu, dialog, popup

UserName

CAS RN 4022-81-5

IUPAC Name: N-[amino(azaniumyl)methylidene]-N''-(4-chlorophenyl)guanidine chloride
Synonyms: N-[amino(ammonio)methylidene]-N''-(4-chlorophenyl)guanidine chloride
Molecular Weight (g/mol): 248.11
Molecular Formula: C8H11Cl2N5
InChi Key: NAFSLMFLGYXGIF-UHFFFAOYSA-N
SMILES: [Cl-].NC([NH3+])=NC(N)=NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results