missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD10529DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CD10529DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

187998-35-2
236.655
OAPQKGGVDCZENK-UHFFFAOYSA-N
2777166
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O
C11H9ClN2O2
MFCD01566260
1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-1-4-chlorophenyl-5-methyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-methyl, buttpark 9618-16, salor-int l317632-1ea, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic-acid, 4-chloro-4-carboxy-5-methyl-1h-pyrazol-1-yl benzene, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-methyl
1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid

Specifications

1-(4-Chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
96.5
97%
C11H9ClN2O2
MFCD01566260
OAPQKGGVDCZENK-UHFFFAOYSA-N
1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
2777166
97%
187998-35-2
100.0
Amber Glass Bottle
5g
1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-1-4-chlorophenyl-5-methyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-methyl, buttpark 9618-16, salor-int l317632-1ea, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic-acid, 4-chloro-4-carboxy-5-methyl-1h-pyrazol-1-yl benzene, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-methyl
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O
236.655
236.66
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents