missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2694342
Change view
Click to view available options
Quantity:
1g
5g
10g
Packaging:
Amber glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD10529DA 1g Amber glass bottle
CD10529DE 5g Amber glass bottle
CD10529EA 10g Amber glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD10529DA Supplier Thermo Scientific Chemicals Supplier No. CD10529DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 187998-35-2
Molecular Formula C11H9ClN2O2
Molecular Weight (g/mol) 236.655
MDL Number MFCD01566260
InChI Key OAPQKGGVDCZENK-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-1-4-chlorophenyl-5-methyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-methyl, buttpark 9618-16, salor-int l317632-1ea, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic-acid, 4-chloro-4-carboxy-5-methyl-1h-pyrazol-1-yl benzene, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-methyl
PubChem CID 2777166
IUPAC Name 1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 236.66
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.