missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic Acid 98.0+%, TCI America™

Supplier:  TCI America C291225G

 View more versions of this product

Catalog No. C291225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic Acid
80789-69-1
25 g
C12H13ClO2
1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O
224.684
224.68
164°C
White
Crystal-Powder at 20°C
MFCD00001373
QJNFJEMGWIQMJT-UHFFFAOYSA-N
1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
97447
≥98.0% (GC,T)

Chemical Identifiers

80789-69-1
224.684
QJNFJEMGWIQMJT-UHFFFAOYSA-N
97447
C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O
C12H13ClO2
MFCD00001373
1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions