missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7125719
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29125G 5 g
C291225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29125G Supplier TCI America Supplier No. C29125G
Only null left

Chemical Identifiers

CAS 80789-69-1
Molecular Formula C12H13ClO2
Molecular Weight (g/mol) 224.684
MDL Number MFCD00001373
InChI Key QJNFJEMGWIQMJT-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
PubChem CID 97447
IUPAC Name 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 164°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 224.68
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic Acid