missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid, 94%, Thermo Scientific™
Chemical Identifiers
| CAS | 50921-39-6 |
|---|---|
| Molecular Formula | C11H11ClO2 |
| Molecular Weight (g/mol) | 210.66 |
| InChI Key | XYSRHOKREWGGFE-UHFFFAOYSA-N |
| Synonym | 1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid |
| PubChem CID | 602761 |
| IUPAC Name | 1-(4-chlorophenyl)cyclobutane-1-carboxylic acid |
| SMILES | C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC170720250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 50921-39-6 | |
| 210.66 | |
| 1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid | |
| 1-(4-chlorophenyl)cyclobutane-1-carboxylic acid |
| C11H11ClO2 | |
| XYSRHOKREWGGFE-UHFFFAOYSA-N | |
| 602761 | |
| C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O |
Specifications
| 50921-39-6 | |
| 92% min. (HPLC) | |
| ClC6H4C4H6CO2H | |
| XYSRHOKREWGGFE-UHFFFAOYSA-N | |
| 1-(4-chlorophenyl)cyclobutane-1-carboxylic acid | |
| 602761 | |
| 94% |
| Authentic | |
| C11H11ClO2 | |
| 1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid | |
| C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O | |
| 210.66 | |
| 210.66 | |
| 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 256-847-3
RUO – Research Use Only