missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid, 94%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 170720250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC170720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

50921-39-6
210.66
1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid
1-(4-chlorophenyl)cyclobutane-1-carboxylic acid
C11H11ClO2
XYSRHOKREWGGFE-UHFFFAOYSA-N
602761
C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
Authentic
Glass bottle
ClC6H4C4H6CO2H
1-4-chlorophenyl cyclobutanecarboxylic acid, 1-4-chlorophenyl-1-cyclobutanecarboxylic acid, 1-4-chlorophenyl cyclobutane-1-carboxylic acid, 1-p-chlorophenyl cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-4-chlorophenyl, acmc-209kr1, 1-4chlorophenyl cyclobutanecarboxylic acid, 1 4-chlorophenyl cyclobutane carboxylic acid, 1-4 chlorophenyl-cyclobutanecarboxylic acid, 1-4-chloro phenyl cyclobutanecarboxylic acid
C1CC(C1)(C2=CC=C(C=C2)Cl)C(=O)O
210.66
210.66
50921-39-6
92% min. (HPLC)
C11H11ClO2
25g
XYSRHOKREWGGFE-UHFFFAOYSA-N
1-(4-chlorophenyl)cyclobutane-1-carboxylic acid
602761
94%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 256-847-3

SDS
Documents

Documents

Promotions

Promotions