missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzhydryl)piperazine 98.0+%, TCI America™

Catalog No. p-7125088
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16111G 1 g
C16115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16111G Supplier TCI America Supplier No. C16111G
Only null left

Chemical Identifiers

CAS 303-26-4
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.803
MDL Number MFCD00005965
InChI Key UZKBSZSTDQSMDR-UHFFFAOYSA-N
Synonym 1-4-chlorobenzhydryl piperazine, norchlorcyclizine, 1-4-chlorophenyl phenyl methyl piperazine, n-p-chlorobenzhydryl piperazine, 4-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl phenylmethyl piperazine, n-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 9340
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

Specifications

Melting Point 75°C
Color White
Boiling Point 180°C
Quantity 1 g
Formula Weight 286.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
RTECSNumber TL1772000
TSCA No