missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzhydryl)piperazine, 90-95%

Catalog No. AC108650250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108650250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC108650250 Supplier Thermo Scientific Chemicals Supplier No. 108650250
Only null left

CAS: 303-26-4 | C17H19ClN2 | 286.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 303-26-4
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.79
InChI Key UZKBSZSTDQSMDR-UHFFFAOYSA-N
Synonym 1-4-chlorobenzhydryl piperazine, norchlorcyclizine, 1-4-chlorophenyl phenyl methyl piperazine, n-p-chlorobenzhydryl piperazine, 4-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl phenylmethyl piperazine, n-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 9340
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

Specifications

Melting Point 65.0°C to 75.0°C
Color White to Beige
Boiling Point 178.0°C to 180.0°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (on dry substance) (Tertiary base)
Packaging Glass bottle
Quantity 25 g
Formula Weight 286.79
Percent Purity 90 to 95%
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 206-137-4

RUO – Research Use Only