missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butylenediphosphonic Acid 98.0+%, TCI America™

Catalog No. B48481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48481G Supplier TCI America Supplier No. B48481G

Chemical Identifiers

CAS 4671-77-6
Molecular Formula C4H8O6P2
Molecular Weight (g/mol) 214.05
MDL Number MFCD01631212
InChI Key JKTORXLUQLQJCM-UHFFFAOYSA-J
Synonym 1,4-butanediphosphonic acid, 1,4-butylenediphosphonic acid, 1,4-diphosphonobutane, tetramethylene phosphonic acid, butane-1,4-bisphosphonic acid, tetramethylenebisphosphonic acid, butane-1,4-diyldiphosphonic acid
PubChem CID 437082
IUPAC Name (4-phosphonatobutyl)phosphonate
SMILES [O-]P([O-])(=O)CCCCP([O-])([O-])=O

Specifications

Melting Point 221°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 218.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Butylenediphosphonic Acid