accessibility menu, dialog, popup

UserName

OPOOPOOO

CAS RN 4671-77-6

IUPAC Name: (4-phosphonatobutyl)phosphonate
Synonyms: 4-phosphonatobutylphosphonate
Molecular Weight (g/mol): 214.05
Molecular Formula: C4H8O6P2
InChi Key: JKTORXLUQLQJCM-UHFFFAOYSA-J
SMILES: [O-]P([O-])(=O)CCCCP([O-])([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results