missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanediol Monomethyl Ether 98.0+%, TCI America™
Supplier: TCI America B240225G
Specifications
| 1,4-Butanediol Monomethyl Ether | |
| Colorless | |
| C5H12O2 | |
| 25 g | |
| KOVAQMSVARJMPH-UHFFFAOYSA-N | |
| 4-methoxybutan-1-ol | |
| 8107 | |
| ≥98.0% (GC) |
| 111-32-0 | |
| 66°C | |
| MFCD00040437 | |
| 4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1 | |
| COCCCCO | |
| 104.15 | |
| 104.15 | |
| Liquid |
Chemical Identifiers
| 111-32-0 | |
| 104.15 | |
| KOVAQMSVARJMPH-UHFFFAOYSA-N | |
| 8107 | |
| COCCCCO |
| C5H12O2 | |
| MFCD00040437 | |
| 4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1 | |
| 4-methoxybutan-1-ol |
Safety and Handling
EINECSNumber : (7)-0097
RTECSNumber : EL4550000
TSCA : No