accessibility menu, dialog, popup

UserName

CAS RN 111-32-0

IUPAC Name: 4-methoxybutan-1-ol
Synonyms: 4-methoxybutan-1-ol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: KOVAQMSVARJMPH-UHFFFAOYSA-N
SMILES: COCCCCO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)