missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol Monomethyl Ether 98.0+%, TCI America™

Catalog No. B24025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24025G 5 g
B240225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24025G Supplier TCI America Supplier No. B24025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-32-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00040437
InChI Key KOVAQMSVARJMPH-UHFFFAOYSA-N
Synonym 4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1
PubChem CID 8107
IUPAC Name 4-methoxybutan-1-ol
SMILES COCCCCO

Specifications

Color Colorless
Boiling Point 66°C
Quantity 5 g
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Butanediol Monomethyl Ether
EINECSNumber (7)-0097
RTECSNumber EL4550000
TSCA No