missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butane diisothiocyanate, 98%

Catalog No. AAL0923403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0923403 1 g
AAL0923406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0923403 Supplier Thermo Scientific Chemicals Supplier No. L0923403

CAS: 4430-51-7 | C6H8N2S2 | 172.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4430-51-7
Molecular Formula C6H8N2S2
Molecular Weight (g/mol) 172.264
MDL Number MFCD00041136
InChI Key RRSISCMPUAGVJN-UHFFFAOYSA-N
Synonym 1,4-butane diisothiocyanate, butane, 1,4-diisothiocyanato, tetramethylene diisothiocyanate, #
PubChem CID 138238
IUPAC Name 1,4-diisothiocyanatobutane
SMILES C(CCN=C=S)CN=C=S

Specifications

Melting Point 30°C to 33°C
Boiling Point 170°C (11 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 1757138
Sensitivity Moisture sensitive
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material 1,4-Butane diisothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only