missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Bromophenyl)pyrrolidine 98.0+%, TCI America™
$70.84 - $234.87
Chemical Identifiers
| CAS | 22090-26-2 |
|---|---|
| Molecular Formula | C10H12BrN |
| Molecular Weight (g/mol) | 226.117 |
| MDL Number | MFCD04112480 |
| InChI Key | BVEGBJXZICCEQW-UHFFFAOYSA-N |
| Synonym | 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene |
| PubChem CID | 7016457 |
| IUPAC Name | 1-(4-bromophenyl)pyrrolidine |
| SMILES | C1CCN(C1)C2=CC=C(C=C2)Br |
Chemical Identifiers
| 22090-26-2 | |
| 226.117 | |
| BVEGBJXZICCEQW-UHFFFAOYSA-N | |
| 7016457 | |
| C1CCN(C1)C2=CC=C(C=C2)Br |
| C10H12BrN | |
| MFCD04112480 | |
| 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene | |
| 1-(4-bromophenyl)pyrrolidine |
Specifications
| 22090-26-2 | |
| White | |
| MFCD04112480 | |
| 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene | |
| C1CCN(C1)C2=CC=C(C=C2)Br | |
| 226.117 | |
| 226.12 | |
| Crystalline Powder |
| 107°C | |
| C10H12BrN | |
| 1 g | |
| BVEGBJXZICCEQW-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)pyrrolidine | |
| 7016457 | |
| ≥98.0% (GC,T) | |
| 1-(4-Bromophenyl)pyrrolidine |
Safety and Handling
TSCA : No