accessibility menu, dialog, popup

UserName

CAS RN 22090-26-2

NBr

CAS RN 22090-26-2

IUPAC Name: 1-(4-bromophenyl)pyrrolidine
Synonyms: 1-(4-bromophenyl)pyrrolidine
Molecular Weight (g/mol): 226.12
Molecular Formula: C10H12BrN
InChi Key: BVEGBJXZICCEQW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1CCCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)