missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC30110EA

 View more versions of this product

Catalog No. CC30110EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-(4-Bromophenyl)pyrrolidine
97%
C10H12BrN
1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene
C1CCN(C1)C2=CC=C(C=C2)Br
226.117
226.12
22090-26-2
Amber Glass Bottle
10g
BVEGBJXZICCEQW-UHFFFAOYSA-N
1-(4-bromophenyl)pyrrolidine
7016457
97%

Chemical Identifiers

22090-26-2
226.117
1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene
1-(4-bromophenyl)pyrrolidine
C10H12BrN
BVEGBJXZICCEQW-UHFFFAOYSA-N
7016457
C1CCN(C1)C2=CC=C(C=C2)Br
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions