missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC30110CB
Specifications
| 1-(4-Bromophenyl)pyrrolidine | |
| 97% | |
| 250mg | |
| BVEGBJXZICCEQW-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)pyrrolidine | |
| 7016457 | |
| 97% |
| 22090-26-2 | |
| C10H12BrN | |
| 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene | |
| C1CCN(C1)C2=CC=C(C=C2)Br | |
| 226.117 | |
| 226.12 |
Chemical Identifiers
| 22090-26-2 | |
| 226.117 | |
| 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene | |
| 1-(4-bromophenyl)pyrrolidine |
| C10H12BrN | |
| BVEGBJXZICCEQW-UHFFFAOYSA-N | |
| 7016457 | |
| C1CCN(C1)C2=CC=C(C=C2)Br |