missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Bromophenyl)imidazole 98.0+%, TCI America™
$147.25 - $721.40
Chemical Identifiers
| CAS | 10040-96-7 |
|---|---|
| Molecular Formula | C9H7BrN2 |
| Molecular Weight (g/mol) | 223.073 |
| MDL Number | MFCD00060489 |
| InChI Key | SERULNRLZWOYPK-UHFFFAOYSA-N |
| PubChem CID | 2735604 |
| IUPAC Name | 1-(4-bromophenyl)imidazole |
| SMILES | C1=CC(=CC=C1N2C=CN=C2)Br |
Chemical Identifiers
| 10040-96-7 | |
| 223.073 | |
| SERULNRLZWOYPK-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)imidazole |
| C9H7BrN2 | |
| MFCD00060489 | |
| 2735604 | |
| C1=CC(=CC=C1N2C=CN=C2)Br |
Specifications
| 10040-96-7 | |
| White | |
| MFCD00060489 | |
| SERULNRLZWOYPK-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)imidazole | |
| 2735604 | |
| ≥98.0% (GC,T) | |
| 1-(4-Bromophenyl)imidazole |
| 123°C | |
| C9H7BrN2 | |
| 1 g | |
| C1=CC(=CC=C1N2C=CN=C2)Br | |
| 223.073 | |
| 223.07 | |
| Crystalline Powder |
Safety and Handling
TSCA : No