accessibility menu, dialog, popup

UserName

NNBr

CAS RN 10040-96-7

IUPAC Name: 1-(4-bromophenyl)-1H-imidazole
Synonyms: 1-(4-bromophenyl)imidazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: SERULNRLZWOYPK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results