missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)imidazole 98.0+%, TCI America™

Catalog No. B41015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41015G 5 g
B41011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41015G Supplier TCI America Supplier No. B41015G
Only null left

Chemical Identifiers

CAS 10040-96-7
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00060489
InChI Key SERULNRLZWOYPK-UHFFFAOYSA-N
PubChem CID 2735604
IUPAC Name 1-(4-bromophenyl)imidazole
SMILES C1=CC(=CC=C1N2C=CN=C2)Br

Specifications

Melting Point 123°C
Color White
Quantity 5 g
Formula Weight 223.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)imidazole