missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)-2-methyl-1-propanone 95.0+%, TCI America™

Catalog No. B52321G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52321G Supplier TCI America Supplier No. B52321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 49660-93-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.101
MDL Number MFCD09891214
InChI Key PMFLKZHWZXWOEH-UHFFFAOYSA-N
PubChem CID 15074783
IUPAC Name 1-(4-bromophenyl)-2-methylpropan-1-one
SMILES CC(C)C(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 87°C
Quantity 1 g
Formula Weight 227.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(4-Bromophenyl)-2-methyl-1-propanone