accessibility menu, dialog, popup

UserName

CAS RN 49660-93-7

IUPAC Name: 1-(4-bromophenyl)-2-methylpropan-1-one
Synonyms: 1-(4-bromophenyl)-2-methylpropan-1-one
Molecular Weight (g/mol): 227.1
Molecular Formula: C10H11BrO
InChi Key: PMFLKZHWZXWOEH-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results