missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 845885-94-1
Molecular Formula C11H10BrN3O2
Molecular Weight (g/mol) 296.124
MDL Number MFCD06659088
InChI Key FJFFHBYCFJNIAV-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid
PubChem CID 2794828
IUPAC Name 1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid
SMILES CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MO07389DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

845885-94-1
296.124
FJFFHBYCFJNIAV-UHFFFAOYSA-N
2794828
CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O
C11H10BrN3O2
MFCD06659088
1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid
1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid

Specifications

845885-94-1
97%
C11H10BrN3O2
1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid
CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O
296.124
296.12
1-(4-Bromobenzyl)-5-methyl-1H-1, 2, 3-triazole-4-carboxylic acid
97.0
Amber Glass Bottle
MFCD06659088
FJFFHBYCFJNIAV-UHFFFAOYSA-N
1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid
2794828
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents