missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. MO07389DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07389DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07389DA Supplier Thermo Scientific Chemicals Supplier No. MO07389DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 845885-94-1
Molecular Formula C11H10BrN3O2
Molecular Weight (g/mol) 296.124
MDL Number MFCD06659088
InChI Key FJFFHBYCFJNIAV-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid
PubChem CID 2794828
IUPAC Name 1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid
SMILES CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 296.12
Percent Purity 97%
Chemical Name or Material 1-(4-Bromobenzyl)-5-methyl-1H-1, 2, 3-triazole-4-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.