missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trimethylsilyl)-1,3-butadiyne, 98%

CAS: 4526-07-2 | C10H18Si2 | 194.42 g/mol

$218.90 - $218.90

Chemical Identifiers

CAS 4526-07-2
Molecular Formula C10H18Si2
Molecular Weight (g/mol) 194.42
MDL Number MFCD00009839
InChI Key LBNVCJHJRYJVPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC#C[Si](C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0924606
View Documents
Thermo Scientific Chemicals
L0924606
5 g N/A
Only null left
 
Description

Description

1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

4526-07-2
194.42
LBNVCJHJRYJVPK-UHFFFAOYSA-N
C10H18Si2
MFCD00009839
C[Si](C)(C)C#CC#C[Si](C)(C)C

Specifications

110°C to 113°C
5 g
MFCD00009839
1758268
Insoluble in water.
C[Si](C)(C)C#CC#C[Si](C)(C)C
194.42
194.43
0.752
4526-07-2
C10H18Si2
UN1325
1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne
LBNVCJHJRYJVPK-UHFFFAOYSA-N
trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane
138279
98%
1,4-Bis(trimethylsilyl)-1,3-butadiyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only