missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trimethylsilyl)-1,3-butadiyne, 98%

Catalog No. aal0924606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0924606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0924606 Supplier Thermo Scientific Chemicals Supplier No. L0924606
Only null left

CAS: 4526-07-2 | C10H18Si2 | 194.42 g/mol

1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 4526-07-2
Molecular Formula C10H18Si2
Molecular Weight (g/mol) 194.42
MDL Number MFCD00009839
InChI Key LBNVCJHJRYJVPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC#C[Si](C)(C)C

Specifications

Melting Point 110°C to 113°C
Quantity 5 g
UN Number UN1325
Beilstein 1758268
Synonym 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne
Solubility Information Insoluble in water.
IUPAC Name trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane
PubChem CID 138279
Formula Weight 194.43
Percent Purity 98%
Density 0.752
Chemical Name or Material 1,4-Bis(trimethylsilyl)-1,3-butadiyne
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only