missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(phenylethynyl)benzene, 97%
CAS: 1849-27-0 | C22H14 | 278.354 g/mol
$87.55 - $824.40
Chemical Identifiers
| CAS | 1849-27-0 |
|---|---|
| Molecular Formula | C22H14 |
| Molecular Weight (g/mol) | 278.354 |
| MDL Number | MFCD00160829 |
| InChI Key | FPVSTPLZJLYNMB-UHFFFAOYSA-N |
| Synonym | 1,4-bis phenylethynyl benzene, 1,4-bis 2-phenylethynyl benzene, benzene,1,4-bis 2-phenylethynyl, p-bis phenylethynyl benzene, 1,4-bis phenylethynyl benzene #, 1,4-bis 2-phenyl-ethynyl-benzene |
| PubChem CID | 624226 |
| IUPAC Name | 1,4-bis(2-phenylethynyl)benzene |
| SMILES | C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3039503
|
Thermo Scientific Chemicals
H3039503 |
1 g |
Each for $87.55
|
|
|||||
|
AAH3039506
|
Thermo Scientific Chemicals
H3039506 |
5 g |
Each for $250.67
|
|
|||||
|
AAH3039514
|
Thermo Scientific Chemicals
H3039514 |
25 g |
Each for $824.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1849-27-0 | |
| 278.354 | |
| FPVSTPLZJLYNMB-UHFFFAOYSA-N | |
| 624226 | |
| C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3 |
| C22H14 | |
| MFCD00160829 | |
| 1,4-bis phenylethynyl benzene, 1,4-bis 2-phenylethynyl benzene, benzene,1,4-bis 2-phenylethynyl, p-bis phenylethynyl benzene, 1,4-bis phenylethynyl benzene #, 1,4-bis 2-phenyl-ethynyl-benzene | |
| 1,4-bis(2-phenylethynyl)benzene |
Specifications
| 1849-27-0 | |
| C22H14 | |
| 1 g | |
| FPVSTPLZJLYNMB-UHFFFAOYSA-N | |
| 1,4-bis(2-phenylethynyl)benzene | |
| 624226 | |
| 97% |
| 176°C to 178°C | |
| MFCD00160829 | |
| 1,4-bis phenylethynyl benzene, 1,4-bis 2-phenylethynyl benzene, benzene,1,4-bis 2-phenylethynyl, p-bis phenylethynyl benzene, 1,4-bis phenylethynyl benzene #, 1,4-bis 2-phenyl-ethynyl-benzene | |
| C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3 | |
| 278.354 | |
| 278.36 | |
| 1,4-Bis(phenylethynyl)benzene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only