CAS RN 1849-27-0
IUPAC Name:
1,4-bis(2-phenylethynyl)benzene
Synonyms:
1,4-bis(2-phenylethynyl)benzene
Molecular Weight (g/mol):
278.35
Molecular Formula:
C22H14
InChi Key:
FPVSTPLZJLYNMB-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C#CC1=CC=C(C=C1)C#CC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1,4-Bis(phenylethynyl)benzene, 97%
1,4-Bis(phenylethynyl)benzene, 97%
CAS: 1849-27-0 Molecular Formula: C22H14 Molecular Weight (g/mol): 278.354 MDL Number: MFCD00160829 InChI Key: FPVSTPLZJLYNMB-UHFFFAOYSA-N Synonym: 1,4-bis phenylethynyl benzene,1,4-bis 2-phenylethynyl benzene,benzene,1,4-bis 2-phenylethynyl,p-bis phenylethynyl benzene,1,4-bis phenylethynyl benzene #,1,4-bis 2-phenyl-ethynyl-benzene PubChem CID: 624226 IUPAC Name: 1,4-bis(2-phenylethynyl)benzene SMILES: C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3