missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(diphenylphosphino)butane 98.0+%, TCI America™

Catalog No. p-7118946
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12465G 5 g
B124625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12465G Supplier TCI America Supplier No. B12465G
Only null left

Chemical Identifiers

CAS 7688-25-7
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00003051
InChI Key BCJVBDBJSMFBRW-UHFFFAOYSA-N
Synonym 1,4-bis diphenylphosphino butane, dppb, phosphine, 1,4-butanediylbis diphenyl, 4-diphenylphosphanyl butyl diphenylphosphane, unii-35hp6ltd2d, tetramethylenebis diphenylphosphine, 35hp6ltd2d, 4-diphenylphosphanylbutyl diphenyl phosphane, 1,4-bis-diphenylphosphino butane
PubChem CID 82124
IUPAC Name [4-(diphenylphosphanyl)butyl]diphenylphosphane
SMILES C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 426.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(diphenylphosphino)butane