missing translation for 'onlineSavingsMsg'
Learn More

[1,4-Bis(diphenylphosphino)butane]palladium(II) Dichloride 96.0+%, TCI America™

Catalog No. B20315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20315G 5 g
B20311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20315G Supplier TCI America Supplier No. B20315G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29964-62-3
Molecular Formula C28H28Cl2P2Pd
Molecular Weight (g/mol) 603.80
MDL Number MFCD02093437
InChI Key JQXJBXVWVPVTOO-UHFFFAOYSA-L
Synonym 1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
PubChem CID 10974057
IUPAC Name [4-(diphenylphosphanyl)butyl]diphenylphosphane; dichloropalladium
SMILES Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 603.80
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,4-Bis(diphenylphosphino)butane]palladium(II) Dichloride