missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(diphenylphosphino)butane, 98%

Catalog No. AC296460050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296460050 5 g Glass bottle
AC296460250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296460050 Supplier Thermo Scientific Chemicals Supplier No. 296460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7688-25-7 | C28H28P2 | 426.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7688-25-7
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00003051
InChI Key BCJVBDBJSMFBRW-UHFFFAOYSA-N
Synonym 1,4-bis diphenylphosphino butane, dppb, phosphine, 1,4-butanediylbis diphenyl, 4-diphenylphosphanyl butyl diphenylphosphane, unii-35hp6ltd2d, tetramethylenebis diphenylphosphine, 35hp6ltd2d, 4-diphenylphosphanylbutyl diphenyl phosphane, 1,4-bis-diphenylphosphino butane
PubChem CID 82124
IUPAC Name 4-diphenylphosphanylbutyl(diphenyl)phosphane
SMILES C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 130.0°C to 137.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1254
Formula Weight 426.47
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Bis(diphenylphosphino)butane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-698-7
TSCA TSCA

RUO – Research Use Only