missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(butyryloxy)-2-butene 95.0+%, TCI America™

Catalog No. p-7123026
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35805G 5 g
B358025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35805G Supplier TCI America Supplier No. B35805G
Only null left

Chemical Identifiers

CAS 1572-84-5
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD07368634
InChI Key OGJKSGMVEHGEKF-UHFFFAOYSA-N
Synonym 2-Butene-1,4-diol Dibutyrate
PubChem CID 53423991
IUPAC Name 4-(butanoyloxy)but-2-en-1-yl butanoate
SMILES CCCC(=O)OCC=CCOC(=O)CCC

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 228.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(butyryloxy)-2-butene