missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%

Catalog No. AAH63961MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63961MD 250 mg
AAH6396103 1 g
AAH6396106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63961MD Supplier Thermo Scientific Chemicals Supplier No. H63961MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 346451-15-8 | C30H30Cl4N2 | 560.384 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 346451-15-8
Molecular Formula C30H30Cl4N2
Molecular Weight (g/mol) 560.384
MDL Number MFCD03844641
InChI Key DLEQNCKNBPYHEY-UHFFFAOYSA-N
Synonym 1,4-bis 4-chlorophenyl phenylmethyl piperazine dihydrochloride, cetirizine impurity d, 1,4-bis 4-chlorophenyl phenylmethyl-piperazine dihydrochloride, 1,4-bis 4-chlorophenyl phenyl methyl piperazine dihydrochloride, 1,4-bis 4-chlorophenyl-phenylmethyl piperazine;dihydrochloride
PubChem CID 24720891
IUPAC Name 1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
SMILES C1CN(CCN1C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl)C(C4=CC=CC=C4)C5=CC=C(C=C5)Cl.Cl.Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 560.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only