missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(acryloyloxy)butane (stabilized with MEHQ) 90.0+%, TCI America™

Catalog No. B2935500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2935500G 500 g
B293525G 25 g
B2935100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B2935500G Supplier TCI America Supplier No. B2935500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1070-70-8
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014940
InChI Key JHWGFJBTMHEZME-UHFFFAOYSA-N
Synonym 1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
PubChem CID 70613
IUPAC Name 4-(prop-2-enoyloxy)butyl prop-2-enoate
SMILES C=CC(=O)OCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 138°C
Quantity 500 g
UN Number 1760
Formula Weight 198.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(acryloyloxy)butane (stabilized with MEHQ)
EINECSNumber (2)-1007&(2)-0958
RTECSNumber UD3130000
TSCA Yes