missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(4-methylstyryl)benzene 98.0+%, TCI America™

Catalog No. B15505G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15505G Supplier TCI America Supplier No. B15505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76439-00-4
Molecular Formula C24H22
Molecular Weight (g/mol) 310.44
MDL Number MFCD00191317
InChI Key BCASZEAAHJEDAL-UHFFFAOYSA-N
PubChem CID 6139005
IUPAC Name 1-methyl-4-(2-{4-[2-(4-methylphenyl)ethenyl]phenyl}ethenyl)benzene
SMILES CC1=CC=C(C=CC2=CC=C(C=CC3=CC=C(C)C=C3)C=C2)C=C1

Specifications

Quantity 5 g
Formula Weight 310.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(4-methylstyryl)benzene