accessibility menu, dialog, popup

UserName

CH3CH3(E/Z)(E/Z)

CAS RN 76439-00-4

IUPAC Name: 1-methyl-4-(2-{4-[2-(4-methylphenyl)ethenyl]phenyl}ethenyl)benzene
Synonyms: 1-methyl-4-(2-{4-[2-(4-methylphenyl)ethenyl]phenyl}ethenyl)benzene
Molecular Weight (g/mol): 310.44
Molecular Formula: C24H22
InChi Key: BCASZEAAHJEDAL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=CC2=CC=C(C=CC3=CC=C(C)C=C3)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results