missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[4-(di-p-tolylamino)styryl]benzene 98.0+%, TCI America™

Catalog No. B20801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20801G Supplier TCI America Supplier No. B20801G

Chemical Identifiers

CAS 55035-43-3
Molecular Formula C50H44N2
Molecular Weight (g/mol) 672.92
MDL Number MFCD12022467
InChI Key LQYYDWJDEVKDGB-XPWSMXQVSA-N
Synonym 1,4-Bis[2-[4-[N,N-di(p-tolyl)amino]phenyl]vinyl]benzene, 4,4′C-[1,4-Phenylenebis(ethene-2,1-diyl)]bis(N,N-di-p-tolylaniline)
PubChem CID 6504535
IUPAC Name N-{4-[(1E)-2-{4-[(1E)-2-{4-[bis(4-methylphenyl)amino]phenyl}ethenyl]phenyl}ethenyl]phenyl}-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(\C=C\C2=CC=C(\C=C\C3=CC=C(C=C3)N(C3=CC=C(C)C=C3)C3=CC=C(C)C=C3)C=C2)C=C1

Specifications

Melting Point 223°C
Quantity 1 g
Formula Weight 672.92
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis[4-(di-p-tolylamino)styryl]benzene