accessibility menu, dialog, popup

UserName

CH3NCH3NH3CCH3(E)(E)

CAS RN 55035-43-3

IUPAC Name: N-{4-[(1E)-2-{4-[(1E)-2-{4-[bis(4-methylphenyl)amino]phenyl}ethenyl]phenyl}ethenyl]phenyl}-4-methyl-N-(4-methylphenyl)aniline
Synonyms: N-{4-[(1E)-2-{4-[(1E)-2-{4-[bis(4-methylphenyl)amino]phenyl}ethenyl]phenyl}ethenyl]phenyl}-4-methyl-N-(4-methylphenyl)aniline
Molecular Weight (g/mol): 672.92
Molecular Formula: C50H44N2
InChi Key: LQYYDWJDEVKDGB-XPWSMXQVSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(\C=C\C2=CC=C(\C=C\C3=CC=C(C=C3)N(C3=CC=C(C)C=C3)C3=CC=C(C)C=C3)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results