missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene 98.0+%, TCI America™

Catalog No. B44285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44285G 5 g
B442825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44285G Supplier TCI America Supplier No. B44285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94525-05-0
Molecular Formula C20H14F6N2O2
Molecular Weight (g/mol) 428.33
MDL Number MFCD07368071
InChI Key LACZRKUWKHQVKS-UHFFFAOYSA-N
PubChem CID 19805071
IUPAC Name 4-{4-[4-amino-2-(trifluoromethyl)phenoxy]phenoxy}-3-(trifluoromethyl)aniline
SMILES NC1=CC(=C(OC2=CC=C(OC3=C(C=C(N)C=C3)C(F)(F)F)C=C2)C=C1)C(F)(F)F

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 428.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene