missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(4,5-dihydro-2-oxazolyl)benzene 98.0+%, TCI America™

Catalog No. B151225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B151225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B151225G Supplier TCI America Supplier No. B151225G
Only null left

Chemical Identifiers

CAS 7426-75-7
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00142545
InChI Key ZDNUPMSZKVCETJ-UHFFFAOYSA-N
Synonym 2,2′C-(1,4-Phenylene)bis(2-oxazoline)
PubChem CID 81929
IUPAC Name 2-[4-(4,5-dihydro-1,3-oxazol-2-yl)phenyl]-4,5-dihydro-1,3-oxazole
SMILES C1CN=C(O1)C1=CC=C(C=C1)C1=NCCO1

Specifications

Melting Point 242°C
Quantity 25 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(4,5-dihydro-2-oxazolyl)benzene
EINECSNumber (5)-3398
TSCA Yes